Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175561.4 | NP_780770.2 | 10 - 11 | 2682 | 68 | |
GenBank mRNA | AK043903.1 | - | 8 - 9 | 2043 | 68 | |
AK220342.1 | - | 2 - 3 | 244 | 68 | ||
BC132334.1 | - | 6 - 7 | 1905 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |