Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008522.3 | NP_032548.2 | 12 - 13 | 1543 | 88 | |
GenBank mRNA | AK036491.1 | - | 12 - 13 | 1544 | 88 | |
AK144556.1 | - | 12 - 13 | 1544 | 88 | ||
AK150799.1 | - | 12 - 13 | 1544 | 88 | ||
AK151822.1 | - | 12 - 13 | 1544 | 88 | ||
AK164781.1 | - | 12 - 13 | 1545 | 88 | ||
BC006904.1 | - | 12 - 13 | 1551 | 3600033 | 88 | |
BC008530.1 | - | 12 - 13 | 1524 | 88 | ||
BC009662.1 | - | 8 - 9 | 974 | 88 | ||
CT010339.1 | - | 12 - 13 | 1509 | 88 | ||
D88510.1 | - | 12 - 13 | 1543 | 88 | ||
FJ538998.1 | - | 13 - 14 | 1509 | 88 | ||
J03298.1 | - | 12 - 13 | 1512 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |