Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138950.2 | NP_620400.2 | 2 - 3 | 4035 | 92 | |
GenBank mRNA | AK131146.1 | - | 2 - 3 | 635 | 92 | |
AK143628.1 | - | 1 - 2 | 123 | 92 | ||
AK160585.1 | - | 1 - 2 | 77 | 92 | ||
AK172019.1 | - | 2 - 3 | 401 | 92 | ||
CN721357.1 | - | 2 - 3 | 545 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |