Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spg7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153176.4 | NP_694816.3 | 7 - 8 | 1005 | 63 | |
GenBank mRNA | AF512565.1 | - | 7 - 8 | 1037 | 63 | |
AF547215.1 | - | 7 - 8 | 1334 | 63 | ||
AK145540.1 | - | 7 - 8 | 1006 | 63 | ||
BC024466.1 | - | 1 - 2 | 66 | 63 | ||
BC024986.1 | - | 5 - 6 | 660 | 63 | ||
BC051051.1 | - | 7 - 8 | 939 | 63 | ||
BC055488.1 | - | 850 | 63 | |||
BC138141.1 | - | 7 - 8 | 1232 | 8861160 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |