Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nae1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144931.3 | NP_659180.1 | 9 - 10 | 758 | 153 | |
GenBank mRNA | AK038756.1 | - | 4 - 5 | 1118 | 153 | |
AK146202.1 | - | 9 - 10 | 716 | 153 | ||
AK162251.1 | - | 9 - 10 | 707 | 153 | ||
BC019163.1 | - | 9 - 10 | 738 | 3711115 | 153 | |
BC021510.1 | - | 9 - 10 | 739 | 5347277 | 153 | |
BC023680.1 | - | 9 - 10 | 740 | 5343341 | 153 | |
BC023897.1 | - | 9 - 10 | 715 | 5321187 | 153 | |
BC027124.1 | - | 9 - 10 | 702 | 5355631 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |