Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198308.1 | NP_938050.1 | 6 - 7 | 863 | 77 | |
GenBank mRNA | AK029575.1 | - | 6 - 7 | 954 | 77 | |
AK129472.1 | - | 6 - 7 | 967 | 77 | ||
AY223867.1 | - | 6 - 7 | 863 | 77 | ||
BC120745.1 | - | 5 - 6 | 793 | 40129668 | 77 | |
BC125425.1 | - | 5 - 6 | 793 | 40129940 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |