Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081241.2 | NP_001074710.2 | 8 - 9 | 778 | 75 | |
GenBank mRNA | AK052316.1 | - | 8 - 9 | 727 | 75 | |
AK122573.1 | - | 8 - 9 | 752 | 75 | ||
AK184855.1 | - | 4 - 5 | 304 | 75 | ||
AK186816.1 | - | 4 - 5 | 303 | 75 | ||
BC079880.1 | - | 8 - 9 | 692 | 6852251 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |