Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009807.2 | NP_033937.2 | 4 - 5 | 646 | 97 | |
GenBank mRNA | BC008152.1 | - | 4 - 5 | 646 | 3583883 | 97 |
L03799.1 | - | 3 - 4 | 448 | 97 | ||
L28095.1 | - | 4 - 5 | 492 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |