Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034878.2 | NP_001030050.2 | 4 - 5 | 849 | 60 | |
GenBank mRNA | BM950889.1 | - | 2 - 3 | 153 | 60 | |
EF565270.1 | - | 3 - 4 | 465 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |