Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166589.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166589.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166590.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166590.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166591.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166591.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166592.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166592.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166593.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166593.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166594.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166594.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166595.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166595.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166596.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_001166596.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_181582.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
NM_181582.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028291.mm.1 |
TC1100003076.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100028297.mm.1 |
NM_001166590.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028297.mm.1 |
NM_001166590.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100028297.mm.1 |
TC1100003076.mm.1 |
MTA TC |