Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wscd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177618.4 | NP_808286.2 | 7 - 8 | 1560 | 60 | |
GenBank mRNA | AK220346.1 | - | 6 - 7 | 1266 | 60 | |
BC030477.1 | - | 7 - 8 | 1697 | 60 | ||
BC048831.1 | - | 6 - 7 | 1379 | 4504495 | 60 | |
BC063070.1 | - | 7 - 8 | 1560 | 5687975 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |