Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163336.1 | NP_001156808.1 | 2 - 3 | 321 | 94 | |
NM_001163337.1 | NP_001156809.1 | 2 - 3 | 321 | 94 | ||
NM_016745.3 | NP_058025.2 | 2 - 3 | 321 | 94 | ||
GenBank mRNA | AK155904.1 | - | 2 - 3 | 322 | 94 | |
BC017639.1 | - | 2 - 3 | 235 | 4924519 | 94 | |
BC026147.1 | - | 2 - 3 | 267 | 4191458 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |