Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016896.3 | NP_058592.1 | 4 - 5 | 622 | 77 | |
GenBank mRNA | AF143094.1 | - | 3 - 4 | 539 | 77 | |
AK036200.1 | - | 4 - 5 | 623 | 77 | ||
AK172518.1 | - | 4 - 5 | 592 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Detect Genomic DNA Mus musculus Probe spans exons |