Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004721.1 | NP_001004721.1 | 4 - 5 | 345 | 85 | |
GenBank mRNA | AK143517.1 | - | 4 - 5 | 345 | 85 | |
AK162913.1 | - | 320 | 85 | |||
AK171988.1 | - | 4 - 5 | 324 | 85 | ||
AK211799.1 | - | 2 - 3 | 125 | 85 | ||
BB839941.1 | - | 4 - 5 | 345 | 85 | ||
BC120680.1 | - | 4 - 5 | 334 | 40129603 | 85 | |
BC120682.1 | - | 4 - 5 | 334 | 40129605 | 85 | |
CK334159.1 | - | 4 - 5 | 298 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |