Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc4ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163452.1 | NP_001156924.1 | 12 - 13 | 1588 | 80 | |
NM_019828.2 | NP_062802.2 | 12 - 13 | 1564 | 80 | ||
GenBank mRNA | AF093589.1 | - | 8 - 9 | 1060 | 80 | |
AF130458.1 | - | 12 - 13 | 1512 | 80 | ||
AF509881.1 | - | 12 - 13 | 1510 | 80 | ||
AK154263.1 | - | 12 - 13 | 1537 | 80 | ||
AK156047.1 | - | 12 - 13 | 1537 | 80 | ||
AK165259.1 | - | 12 - 13 | 1537 | 80 | ||
AK170085.1 | - | 12 - 13 | 1526 | 80 | ||
AK171404.1 | - | 12 - 13 | 1537 | 80 | ||
AK182119.1 | - | 2 - 3 | 162 | 80 | ||
AK192661.1 | - | 2 - 3 | 161 | 80 | ||
AK194397.1 | - | 2 - 3 | 163 | 80 | ||
AK195086.1 | - | 2 - 3 | 162 | 80 | ||
AK195280.1 | - | 182 | 80 | |||
AK220239.1 | - | 12 - 13 | 1564 | 80 | ||
BC003931.1 | - | 5 - 6 | 524 | 80 | ||
BC033274.1 | - | 12 - 13 | 1516 | 3673725 | 80 | |
BC057330.1 | - | 12 - 13 | 1546 | 6816155 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |