Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027303.1 | NP_081579.1 | 3 - 4 | 652 | 114 | |
GenBank mRNA | AK006546.1 | - | 3 - 4 | 594 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons |