Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019707.4 | NP_062681.2 | 2 - 3 | 255 | 67 | |
GenBank mRNA | AB022100.1 | - | 2 - 3 | 264 | 67 | |
AK016527.1 | - | 3 - 4 | 1513 | 67 | ||
AK029195.1 | - | 2 - 3 | 214 | 67 | ||
AK032409.1 | - | 2 - 3 | 284 | 67 | ||
AK034995.1 | - | 2 - 3 | 235 | 67 | ||
AK039438.1 | - | 2 - 3 | 277 | 67 | ||
AK045201.1 | - | 2 - 3 | 256 | 67 | ||
AK048724.1 | - | 2 - 3 | 255 | 67 | ||
AK085939.1 | - | 2 - 3 | 215 | 67 | ||
AK134649.1 | - | 2 - 3 | 214 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |