Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011521.2 | NP_035651.1 | 2 - 3 | 224 | 79 | |
GenBank mRNA | AK134326.1 | - | 2 - 3 | 222 | 79 | |
AK145805.1 | - | 2 - 3 | 223 | 79 | ||
AK153445.1 | - | 2 - 3 | 224 | 79 | ||
AK159215.1 | - | 2 - 3 | 223 | 79 | ||
AK164952.1 | - | 2 - 3 | 223 | 79 | ||
BC005679.1 | - | 2 - 3 | 209 | 3154160 | 79 | |
D89571.1 | - | 2 - 3 | 224 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |