Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277188.1 | NP_001264117.1 | 15 - 16 | 1890 | 95 | |
NR_102339.1 | 14 - 15 | 1847 | 95 | |||
GenBank mRNA | BC065114.1 | - | 14 - 15 | 1847 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |