Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277188.1 | NP_001264117.1 | 25 - 26 | 3065 | 65 | |
NR_102339.1 | 24 - 25 | 3022 | 65 | |||
GenBank mRNA | BC065114.1 | - | 24 - 25 | 3022 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |