Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178743.3 | NP_848858.2 | 4 - 5 | 587 | 80 | |
GenBank mRNA | AF345196.1 | - | 4 - 5 | 587 | 80 | |
AK078545.1 | - | 4 - 5 | 564 | 80 | ||
AK089182.1 | - | 4 - 5 | 567 | 80 | ||
BC038604.1 | - | 4 - 5 | 580 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |