Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028710.3 | NP_082986.3 | 10 - 11 | 1843 | 60 | |
GenBank mRNA | AK018132.2 | - | 7 - 8 | 1260 | 60 | |
AK158726.1 | - | 10 - 11 | 1844 | 60 | ||
AK173082.1 | - | 7 - 8 | 942 | 60 | ||
AK186315.1 | - | 2 - 3 | 60 | |||
AK192114.1 | - | 2 - 3 | 249 | 60 | ||
BC022158.1 | - | 6 - 7 | 673 | 60 | ||
BC084731.1 | - | 7 - 8 | 1533 | 6813261 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |