Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Necab3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021546.3 | NP_067521.2 | 8 - 9 | 708 | 65 | |
GenBank mRNA | AB039948.1 | - | 6 - 7 | 529 | 65 | |
AK044296.1 | - | 8 - 9 | 700 | 65 | ||
AK158225.1 | - | 8 - 9 | 710 | 65 | ||
BC126874.1 | - | 6 - 7 | 522 | 40044848 | 65 | |
BC126875.1 | - | 6 - 7 | 522 | 40044849 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |