Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Endov | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164636.1 | NP_001158108.1 | 3 - 4 | 421 | 61 | |
NM_177394.3 | NP_796368.1 | 2 - 3 | 473 | 61 | ||
GenBank mRNA | AK042626.1 | - | 2 - 3 | 361 | 61 | |
AK084233.1 | - | 3 - 4 | 422 | 61 | ||
AK084548.1 | - | 3 - 4 | 297 | 61 | ||
AK170595.1 | - | 2 - 3 | 466 | 61 | ||
AK172181.1 | - | 2 - 3 | 440 | 61 | ||
AK179832.1 | - | 1 - 2 | 118 | 61 | ||
BC056486.1 | - | 2 - 3 | 393 | 5697870 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |