Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herpud1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022331.1 | NP_071726.1 | 1 - 2 | 237 | 76 | |
GenBank mRNA | AB034991.1 | - | 1 - 2 | 237 | 76 | |
AK081688.1 | - | 1 - 2 | 244 | 76 | ||
AK159500.1 | - | 1 - 2 | 243 | 76 | ||
AK165832.1 | - | 1 - 2 | 243 | 76 | ||
AK172874.1 | - | 1 - 2 | 230 | 76 | ||
BC013523.1 | - | 1 - 2 | 229 | 4236803 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |