Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009764.3 | NP_033894.3 | 17 - 18 | 5183 | 114 | |
GenBank mRNA | AB221610.1 | - | 8 - 9 | 2412 | 114 | |
BC003248.1 | - | 3 - 4 | 391 | 114 | ||
BC068303.1 | - | 17 - 18 | 5016 | 30431022 | 114 | |
U31625.1 | - | 17 - 18 | 5040 | 114 | ||
U32446.1 | - | 17 - 18 | 5093 | 114 | ||
U36475.1 | - | 16 - 17 | 5003 | 114 | ||
U68174.1 | - | 17 - 18 | 5087 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons |