Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myh7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085378.2 | NP_001078847.1 | 16 - 17 | 1680 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |