Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028838.2 | NP_083114.2 | 7 - 8 | 1082 | 105 | |
GenBank mRNA | AJ428068.1 | - | 7 - 8 | 1081 | 105 | |
AK010252.1 | - | 7 - 8 | 1081 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Recommended Assay Mus musculus Inventoried Probe spans exons |