Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800012223.mm.1 |
NM_007662.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012223.mm.1 |
NM_007662.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012223.mm.1 |
TC0800001501.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012226.mm.1 |
NM_007662.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012226.mm.1 |
NM_007662.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012226.mm.1 |
TC0800001501.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012225.mm.1 |
NM_007662.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012225.mm.1 |
NM_007662.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012225.mm.1 |
TC0800001501.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012224.mm.1 |
NM_007662.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012224.mm.1 |
NM_007662.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012224.mm.1 |
TC0800001501.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800012227.mm.1 |
NM_007662.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012227.mm.1 |
NM_007662.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800012227.mm.1 |
TC0800001501.mm.1 |
MTA TC |