Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136062.2 | NP_001129534.1 | 3 - 4 | 277 | 71 | |
NM_001276285.1 | NP_001263214.1 | 3 - 4 | 289 | 71 | ||
NM_007933.3 | NP_031959.1 | 3 - 4 | 389 | 71 | ||
GenBank mRNA | AK002485.1 | - | 3 - 4 | 249 | 71 | |
BC013460.1 | - | 2 - 3 | 261 | 4217678 | 71 | |
BY115250.1 | - | 3 - 4 | 262 | 71 | ||
CT010193.1 | - | 2 - 3 | 183 | 71 | ||
X57747.1 | - | 3 - 4 | 240 | 71 | ||
X62667.1 | - | 2 - 3 | 233 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |