Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prmt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145404.1 | NP_663379.1 | 4 - 5 | 411 | 82 | |
GenBank mRNA | AK154255.1 | - | 4 - 5 | 438 | 82 | |
AK173304.1 | - | 4 - 5 | 431 | 82 | ||
AK182211.1 | - | 1 - 2 | 106 | 82 | ||
AY673972.1 | - | 3 - 4 | 281 | 82 | ||
BC006705.1 | - | 4 - 5 | 411 | 3583763 | 82 | |
BC057177.1 | - | 4 - 5 | 435 | 5010547 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |