Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172674.2 | NP_766262.2 | 1 - 2 | 148 | 80 | |
GenBank mRNA | AK038573.1 | - | 1 - 2 | 55 | 80 | |
AK045309.1 | - | 1 - 2 | 67 | 80 | ||
CJ176261.1 | - | 1 - 2 | 149 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Inventoried Probe spans exons |