Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ergic3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025516.4 | NP_079792.1 | 5 - 6 | 475 | 56 | |
GenBank mRNA | AK009350.1 | - | 5 - 6 | 476 | 56 | |
AK013942.1 | - | 5 - 6 | 475 | 56 | ||
AK050285.1 | - | 5 - 6 | 490 | 56 | ||
AK190333.1 | - | 1 - 2 | 51 | 56 | ||
BC043720.1 | - | 5 - 6 | 500 | 5375947 | 56 | |
BC057130.1 | - | 5 - 6 | 494 | 6819420 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |