Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cirh1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011574.2 | NP_035704.2 | 2 - 3 | 234 | 78 | |
GenBank mRNA | AK144455.1 | - | 2 - 3 | 219 | 78 | |
AK146263.1 | - | 2 - 3 | 210 | 78 | ||
AK150862.1 | - | 2 - 3 | 78 | |||
AK168470.1 | - | 2 - 3 | 184 | 78 | ||
AK173318.1 | - | 1 - 2 | 175 | 78 | ||
BC068113.1 | - | 2 - 3 | 223 | 5707865 | 78 | |
CN718589.1 | - | 2 - 3 | 234 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |