Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grin2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010350.2 | NP_034480.2 | 13 - 14 | 3125 | 57 | |
GenBank mRNA | AK147390.1 | - | 11 - 12 | 3182 | 57 | |
AK165770.1 | - | 9 - 10 | 2309 | 57 | ||
BC137849.1 | - | 11 - 12 | 2515 | 8860868 | 57 | |
D10694.1 | - | 10 - 11 | 2352 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |