Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131020.1 | NP_001124492.1 | 2 - 3 | 593 | 92 | |
NM_010277.3 | NP_034407.2 | 2 - 3 | 593 | 92 | ||
GenBank mRNA | AF332061.1 | - | 2 - 3 | 420 | 92 | |
AF332062.1 | - | 2 - 3 | 420 | 92 | ||
AK079577.1 | - | 2 - 3 | 527 | 92 | ||
AK140151.1 | - | 2 - 3 | 525 | 92 | ||
BC100728.1 | - | 2 - 3 | 472 | 92 | ||
BC100737.1 | - | 2 - 3 | 465 | 92 | ||
BC101968.1 | - | 2 - 3 | 472 | 92 | ||
BC103571.1 | - | 2 - 3 | 466 | 92 | ||
BC139357.1 | - | 2 - 3 | 593 | 8862379 | 92 | |
BC139358.1 | - | 2 - 3 | 593 | 8862380 | 92 | |
EF101554.1 | - | 2 - 3 | 239 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |