Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015751.2 | NP_056566.2 | 16 - 17 | 1860 | 83 | |
GenBank mRNA | AK049299.1 | - | 16 - 17 | 1862 | 83 | |
AK134738.1 | - | 16 - 17 | 1797 | 83 | ||
AK144711.1 | - | 16 - 17 | 1792 | 83 | ||
AK147151.1 | - | 16 - 17 | 1798 | 83 | ||
AK167130.1 | - | 16 - 17 | 1808 | 83 | ||
AK167370.1 | - | 16 - 17 | 1798 | 83 | ||
AK167704.1 | - | 16 - 17 | 1759 | 83 | ||
BC005422.1 | - | 16 - 17 | 1781 | 3489529 | 83 | |
BC066794.1 | - | 15 - 16 | 1563 | 83 | ||
BC066836.1 | - | 15 - 16 | 1563 | 83 | ||
U90446.1 | - | 16 - 17 | 1749 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |