Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim67 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198632.2 | NP_941034.2 | 7 - 8 | 2658 | 80 | |
GenBank mRNA | BC094596.1 | - | 7 - 8 | 2585 | 5705515 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |