Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199015.1 | NP_001185944.1 | 11 - 12 | 1286 | 95 | |
NM_001199016.1 | NP_001185945.1 | 10 - 11 | 1174 | 95 | ||
NM_021291.3 | NP_067266.1 | 10 - 11 | 1401 | 95 | ||
GenBank mRNA | AF192310.1 | - | 11 - 12 | 1286 | 95 | |
AJ249198.1 | - | 10 - 11 | 1401 | 95 | ||
AK142541.1 | - | 10 - 11 | 1175 | 95 | ||
AK164714.1 | - | 9 - 10 | 1591 | 95 | ||
BC010746.1 | - | 11 - 12 | 1264 | 4236578 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |