Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400025181.mm.1 |
TC0400003171.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025181.mm.1 |
NM_146254.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025181.mm.1 |
NM_146254.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025184.mm.1 |
TC0400003171.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025184.mm.1 |
NM_146254.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025184.mm.1 |
NM_146254.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025180.mm.1 |
TC0400003171.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025180.mm.1 |
NM_146254.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025180.mm.1 |
NM_146254.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025182.mm.1 |
TC0400003171.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400025182.mm.1 |
NM_146254.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400025182.mm.1 |
NM_146254.4 |
Refseq |