Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039081.2 | NP_001034170.1 | 5 - 6 | 578 | 115 | |
NM_001286221.1 | NP_001273150.1 | 7 - 8 | 709 | 115 | ||
NM_001286222.1 | NP_001273151.1 | 7 - 8 | 763 | 115 | ||
NM_001286223.1 | NP_001273152.1 | 5 - 6 | 559 | 115 | ||
NM_027696.3 | NP_081972.2 | 7 - 8 | 766 | 115 | ||
GenBank mRNA | AK129405.1 | - | 7 - 8 | 686 | 115 | |
AK168693.1 | - | 5 - 6 | 559 | 115 | ||
AY769435.1 | - | 6 - 7 | 558 | 115 | ||
AY769436.1 | - | 5 - 6 | 481 | 115 | ||
AY769437.1 | - | 5 - 6 | 531 | 115 | ||
AY769438.1 | - | 7 - 8 | 631 | 115 | ||
BC043930.1 | - | 7 - 8 | 691 | 115 | ||
BC108273.1 | - | 5 - 6 | 545 | 30080243 | 115 | |
BC130227.1 | - | 5 - 6 | 530 | 40141863 | 115 | |
BQ562973.1 | - | 2 - 3 | 156 | 115 | ||
CN538186.1 | - | 2 - 3 | 179 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |