Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Habp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146101.1 | NP_666213.1 | 2 - 3 | 150 | 75 | |
GenBank mRNA | AK132407.1 | - | 167 | 75 | ||
BC031775.1 | - | 2 - 3 | 150 | 4242577 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |