Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actr10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019785.2 | NP_062759.2 | 9 - 10 | 817 | 139 | |
GenBank mRNA | AF190797.1 | - | 9 - 10 | 784 | 139 | |
AK090043.1 | - | 9 - 10 | 821 | 139 | ||
AK161642.1 | - | 9 - 10 | 777 | 139 | ||
BC002227.1 | - | 9 - 10 | 777 | 3491299 | 139 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |