Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018868.2 | NP_061356.2 | 11 - 12 | 1377 | 75 | |
GenBank mRNA | AF053232.1 | - | 11 - 12 | 1282 | 75 | |
AK044216.1 | - | 1374 | 75 | |||
AK133959.1 | - | 11 - 12 | 1375 | 75 | ||
AK134286.1 | - | 11 - 12 | 1378 | 75 | ||
BC076604.1 | - | 11 - 12 | 1378 | 30630088 | 75 | |
BC085135.1 | - | 11 - 12 | 1347 | 5718488 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |