Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ica1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027407.3 | NP_081683.3 | 8 - 9 | 1164 | 94 | |
GenBank mRNA | AK006525.1 | - | 6 - 7 | 692 | 94 | |
AK029991.1 | - | 8 - 9 | 936 | 94 | ||
AK158859.1 | - | 8 - 9 | 1000 | 94 | ||
BC058983.1 | - | 3 - 4 | 293 | 94 | ||
BC079669.1 | - | 8 - 9 | 1164 | 30355976 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |