Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042634.2 | NP_001036099.1 | 11 - 12 | 1384 | 112 | |
NR_027853.1 | 10 - 11 | 1293 | 112 | |||
NR_027854.1 | 9 - 10 | 2642 | 112 | |||
GenBank mRNA | AK076199.1 | - | 11 - 12 | 1319 | 112 | |
AK135765.1 | - | 11 - 12 | 1319 | 112 | ||
AK146091.1 | - | 5 - 6 | 1674 | 112 | ||
BC062887.1 | - | 2594 | 112 | |||
L29221.1 | - | 10 - 11 | 1209 | 112 | ||
M38381.1 | - | 11 - 12 | 1300 | 112 | ||
X57186.1 | - | 11 - 12 | 1284 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |