Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aox4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023631.2 | NP_076120.2 | 1 - 2 | 239 | 123 | |
GenBank mRNA | AF233581.1 | - | 1 - 2 | 166 | 123 | |
AK158427.1 | - | 1 - 2 | 240 | 123 | ||
AK160953.1 | - | 1 - 2 | 205 | 123 | ||
BC117975.1 | - | 1 - 2 | 166 | 40098789 | 123 | |
BC117976.1 | - | 1 - 2 | 166 | 40098791 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |