Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008514.4 | NP_032540.2 | 17 - 18 | 3951 | 67 | |
GenBank mRNA | AF074265.1 | - | 17 - 18 | 3820 | 67 | |
AK140796.1 | - | 5 - 6 | 1346 | 67 | ||
AK162203.1 | - | 4 - 5 | 755 | 67 | ||
BC060704.1 | - | 17 - 18 | 3951 | 6409058 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |