Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mpp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164682.1 | NP_001158154.1 | 20 - 21 | 2227 | 65 | |
NM_145143.3 | NP_660125.2 | 19 - 20 | 1590 | 65 | ||
GenBank mRNA | AB059357.1 | - | 18 - 19 | 1481 | 65 | |
AB059358.1 | - | 17 - 18 | 1463 | 65 | ||
AJ748820.1 | - | 18 - 19 | 1471 | 65 | ||
AK044682.1 | - | 20 - 21 | 2226 | 65 | ||
BC058776.1 | - | 19 - 20 | 3094 | 65 | ||
BC061694.1 | - | 19 - 20 | 1507 | 6468172 | 65 | |
BC116723.1 | - | 14 - 15 | 1110 | 40107532 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |