Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172406.3 | NP_765994.2 | 8 - 9 | 1344 | 71 | |
GenBank mRNA | AK014647.1 | - | 8 - 9 | 1293 | 71 | |
AK036650.1 | - | 8 - 9 | 1293 | 71 | ||
BC060681.1 | - | 7 - 8 | 1061 | 6410345 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |